Anti-TORC2 Ser171 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-3415R-A750
BOSSBS-3415R-A750EA 6850 DKK
BOSSBS-3415R-A750
Anti-TORC2 Ser171 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites. Acts as a coactivator, in the SIK/TORC signaling pathway, being active when dephosphorylated and acts independently of CREB1 'Ser-133' phosphorylation. Enhances the interaction of CREB1 with TAF4. Regulates gluconeogenesis as a component of the LKB1/AMPK/TORC2 signaling pathway. Regulates the expression of specific genes such as the steroidogenic gene, StAR. Potent coactivator of PPARGC1A and inducer of mitochondrial biogenesis in muscle cells. AlsoMore Product Information