Anti-RNF135 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-11753R-HRP
BOSSBS-11753R-HRPEA 6720 DKK
BOSSBS-11753R-HRP
Anti-RNF135 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF135 (RING finger protein 135), also known as L13, is a 432 amino acid protein that contains one RING-type zinc finger and one SPRY domain. Via its RING-type zinc finger, RNF135 may play a role in transcriptional regulation and protein degradation events. Defects in the gene encoding RNF135 are theMore Product Information