Anti-CALY Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-11719R-CY7
BOSSBS-11719R-CY7EA 6720 DKK
BOSSBS-11719R-CY7
Anti-CALY Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Calcyon is a single transmembrane protein that interacts with D1 dopamine receptors. Dopamine is a neurotransmitter that regulates synaptic transmission involved in learning and memory. D1 receptors, the most abundant dopamine receptor in the central nervous system, appear to modulate the activity of D2 dopamine receptors, mediate various behavioural responses, and regulate neuron growth and differentiation. Calcyon is present in neuronal cell bodies and processes of the cortex and hippocampus, and it is especially abundant in pyramidal neurons. Interaction of Calcyon with D1 receptorsMore Product Information