Anti-ABLIM1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-11449R-FITC
BOSSBS-11449R-FITCEA 6720 DKK
BOSSBS-11449R-FITC
Anti-ABLIM1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
The C. elegans protein Unc-115 mediates axon guidance by modulating the growth cone Actin cytoskeleton in response to signals received by growth cone receptors. The mammalian homolog of Unc-115 is the Actin-binding LIM protein family member 1 (ABLIM1, also designated Limatin). The ABLIM1 protein has an N-terminal domain that contains four double zinc finger motifs, which conform to the LIM motif consensus sequence. ABLIM1 binds to F-Actin through a dematin-like domain and is expressed in retina, brain and muscle tissue. There are four known isoforms of ABLIM1. The gene encoding ABLIM1 mapsMore Product Information