Anti-HAPLN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-11051R-A555
BOSSBS-11051R-A555EA 6670 DKK
BOSSBS-11051R-A555
Anti-HAPLN2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
HAPLN2 is a 340 amino acid protein encoded by the human gene HAPLN2. HAPLN2 belongs to the HAPLN family and contains one immunoglobulin (Ig)-like, V-type domain and two link domains. HAPLN2 mediates a firm binding of versican V2 to hyaluronic acid. HAPLN2 is believed to play a pivotal role in the formation of the hyaluronan-associated matrix in the central nervous system (CNS), which facilitates neuronal conduction and general structural stabilization. HAPLN2 may also be involved in the formation of extracellular matrices, contributing to perineuronal nets and facilitating the understandingMore Product Information