Anti-MUC16 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-0091R-A647
BOSSBS-0091R-A647EA 6730 DKK
BOSSBS-0091R-A647
Anti-MUC16 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR