Desiccator cabinet, SICCO Super Big Star
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
V1935-07
BOHLV1935-07EA 26350 DKK
BOHLV1935-07
Desiccator cabinet, SICCO Super Big Star
Ekssikatorer Ekssikatorkabinetter
SICCO desiccators are designed for storing and drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination in the atmosphere.
  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Non slip rubber feet
  • Easy to read electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Variable height shelves made of stainless steel
  • Telescopic shelves
  • Temperature resistance: –20 to +70 °C
The controlled environment inside the desiccator makesMore Product Information