Desiccator cabinet, SICCO Star Thermo
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
V1830-07
BOHLV1830-07EA 58350 DKK
BOHLV1830-07
Desiccator cabinet, SICCO Star Thermo
Ekssikatorer Ekssikatorkabinetter
Aluminium frame with 2-pane insulation (inside glass, outside acrylic glass), including four shelves made of PC, usable with up to max. 26 shelves. Integrated heating (850 W) with temperature sensor and air circulation. Electrical power connection 230 V/50 Hz is required.
  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Easy to read electronic hygrometer
  • Variable adjustable shelves
  • Extractable shelves
  • Non-slip rubber feet
  • UVC 1
SICCO Desiccators are designed for storing or drying humidity sensitiveMore Product Information