Tubing cutter, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S1852-28
BOHLS1852-28EA 841 DKK
BOHLS1852-28
Tubing cutter, BOLA
Knive
Ideal for cutting plastic and rubber tubing with and without textile reinforcement.
  • Blade is exchangeable
  • Not suitable for steel reinforced tubing