Microplate incubator shaker, Jitterbug™
Leverandør: BOEKEL SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
130000
BOEI130000EA 42550 DKK
BOEI130000
Microplate incubator shaker, Jitterbug™
Rystere Mikropladerystere
Quiet, rugged shaker is ideal for a variety of immunoassay and research applications.
Accommodates two microplates. Microprocessor-based for independent setting of temperature, shaking speed, and mixing duration. The warmed inner aluminium bed provides temperature uniformity. An inner chamber cover reduces the risk of biohazard from the disbursement of aerosols and insulates the plates during incubation. Outer tinted Plexiglas® cover further insulates and protects sensitive reagents from light. A circulating fan with auto shutoff prevents condensation. Timed operation from 1 second to 99More Product Information