Forceps
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1800 1801 1802
232-0013EA 332 DKK
232-0013 BOCH1801 232-0015
Forceps
Pincet
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very high chemical and thermal resistance
  • Smooth surface prevents any contamination and makes cleaning easier