Wire baskets, square and rectangular
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10023 10071 10073 10076 10029 10025 10026 10028 10030 10031 10032 10033 10027 10020 10021 10022 10024
BOCH10023EA 597 DKK
BOCH10023 BOCH10071 BOCH10073 BOCH10076 BOCH10029 BOCH10025 BOCH10026 BOCH10028 BOCH10030 BOCH10031 BOCH10032 BOCH10033 BOCH10027 BOCH10020 BOCH10021 BOCH10022 BOCH10024
Wire baskets, square and rectangular
Kurve Kurve, tråd
These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Very good chemical and temperature resistance