Anti-CD37 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: IPO-24]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNUM0170-50
BNUM0170-50EA 3110 DKK
BNUM0170-50
Anti-CD37 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: IPO-24]
Antistoffer
Recognizes a protein of 33-55 kDa, identified as CD37 (Workshop V; Code CD37.7). CD37 is strongly expressed on normal and neoplastic mature (sIg ) B-lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells.1 CD37 is also present, at low densities, on resting and activated T cells, neutrophils, monocytes, and some myelomonocytic leukemia cells. It is absent from platelets, erythrocytes. CD37 is a member of a family of tetraspan transmembrane proteins, including CD9,More Product Information