Anti-CD44 Mouse Monoclonal Antibody (Purified) [clone: DF1485]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNUB1037-500 BNUB1037-100
BNUB1037-500EA 3810 DKK
BNUB1037-500 BNUB1037-100
Anti-CD44 Mouse Monoclonal Antibody (Purified) [clone: DF1485]
Antistoffer
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cellMore Product Information