Fernbach culture flasks, with DeLong® style neck
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2550-02800 2554-02800 2554-01800 2554-02833
BELC2550-02800EA 3390.38 DKK
BELC2550-02800 BELC2554-02800 BELC2554-01800 BELC2554-02833
Fernbach culture flasks, with DeLong® style neck
Kolber Dyrkningsbeholdere
Manufactured from type 1 class A borosilicate glass, these flasks are designed for culturing organisms requiring a large surface area-to-volume ratio.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes