Culture shake flasks, with baffles and DeLong® style neck
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2543-00125 2543-00300 2543-00500 2543-01000 2543-02000 2543-00250 2543-03000 2543-61000 2543-62000 2543-64000 2543-04000 2547-00500 2543-60250 2543-60500 2547-00250 2543-00050
BELC2543-00125EA 2280.91 DKK
BELC2543-00125 BELC2543-00300 BELC2543-00500 BELC2543-01000 BELC2543-02000 BELC2543-00250 BELC2543-03000 BELC2543-61000 BELC2543-62000 BELC2543-64000 BELC2543-04000 BELC2547-00500 BELC2543-60250 BELC2543-60500 BELC2547-00250 BELC2543-00050
Culture shake flasks, with baffles and DeLong® style neck
Kolber Dyrkningsbeholdere
Made of type 1 class A borosilicate glass, these shake flasks are the basic tool for growth studies of aerobic microbial cultures and have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation.
  • Very good chemical resistance
  • Very good temperature resistance
Designed for use with orbital shakers.