Culture shake flasks
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2541-00125 2541-00250 2541-01000 2541-02000 2542-00500 2542-01000 2542-02000 2542-00250
BELC2541-00125EA 2001.38 DKK
BELC2541-00125 BELC2541-00250 BELC2541-01000 BELC2541-02000 BELC2542-00500 BELC2542-01000 BELC2542-02000 BELC2542-00250
Culture shake flasks
Kolber Dyrkningsbeholdere
Flasks, made of type 1 class A borosilicate glass, with rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation.
  • With break resistant lip to resist chipping
  • Very good chemical resistance
  • Very good temperature resistance
Designed for use with orbital shakers.