Disposable caps for culture tubes and flasks, KAP-UTS™
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2007-13001 2007-16001 2007-16002 2007-16005 2007-18005 2007-25001 2007-25002 2007-25003 2007-25004 2007-25005 2007-25006 2007-25013 2007-13002 2007-13005 2007-18002 2007-13003 2007-13004 2007-13006 2007-16003 2007-16006 2007-18006 2007-20004 2007-38005 2007-20002 2007-20006
BELC2007-13001EA 2125.21 DKK
BELC2007-13001 BELC2007-16001 BELC2007-16002 BELC2007-16005 BELC2007-18005 BELC2007-25001 BELC2007-25002 BELC2007-25003 BELC2007-25004 BELC2007-25005 BELC2007-25006 BELC2007-25013 BELC2007-13002 BELC2007-13005 BELC2007-18002 BELC2007-13003 BELC2007-13004 BELC2007-13006 BELC2007-16003 BELC2007-16006 BELC2007-18006 BELC2007-20004 BELC2007-38005 BELC2007-20002 BELC2007-20006
Disposable caps for culture tubes and flasks, KAP-UTS™
Låg Reagensglaslåg
Made of nontoxic polypropylene.
  • Autoclavable to 140 ºC
  • Same evaporation and sterility characteristics as stainless steel closures
  • Molded pressure fins for firm grip
Designed for culture tubes and flasks with DeLong® style necks and access ports.