Serological pipettes, Shortie
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1224-05010
BELC1224-05010EA 1308.77 DKK
BELC1224-05010
Serological pipettes, Shortie
Pipetter Serologiske pipetter
These serological pipettes are manufactured from type 1 borosilicate glass.
  • Shorter length facilitates use under biological hoods
  • Plain top promotes rapid, even flow (use rubber bulb to fill)
  • Calibrated to tip
  • Tempered tip; to deliver, blow-out
  • Will withstand repeated autoclaving