Vibration damping mounts, Vibrasorb®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H183860001
BELAH183860001EA 22140 DKK
BELAH183860001
Vibration damping mounts, Vibrasorb®, SP Bel-Art
Møbler Borde
Vibrasorb® vibration damping mounts facilitate optimal use and help protect electromechanical and optical equipment from excessive wear and even structural damage that can be caused by exposure to vibration.
  • Polished surface resists scratches and chemical attack
  • Thick platforms (64 mm) moulded of dense black and white terrazzo
Larger size will absorb vibrations down to 13 Hz with a 16 kg load whereas smaller size will absorb vibrations down to 16 Hz with a 16 kg.
Leveringsinformation: Supplied with four Vibro-Absorber™ cushions with neoprene feetMore Product Information