Supports for round bottom flasks, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F389500000
BELAF389500000EA 945 DKK
BELAF389500000
Supports for round bottom flasks, SP Bel-Art
Beslag Flaskeholdere
This PP round bottom flask support fits any size round-bottom flask up to 10 liters using stepped, concentric rings.
  • Stackable
  • Hands-free, upright position
  • Good chemical and temperature resistance
Use several for adjustable work height.