Forceps, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F379200000
BELAF379200000EA 276 DKK
BELAF379200000
Forceps, SP Bel-Art
Pincet
PP, white.
  • Strong and durable
  • Serrated tips ensure a firm grasp
  • Autoclavable up to 121 °C