Mini cryocooler, –15 °C, Bel-Art ProCulture, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F188460020
BELAF188460020EA 2800 DKK
BELAF188460020
Mini cryocooler, –15 °C, Bel-Art ProCulture, SP Bel-Art
Bokse Kølere/Frysebokse/Poser
Polycarbonate cooler body gel-filled with clear polycarbonate lid.
  • Adapters change well size to stow 0,5 ml microcentrifuge tubes
  • Small size is ideal for benchtop use
  • All polycarbonate cooler body cleans easily
  • Use to protect samples in freezers with defrost cycle and fluctuating temperatures
A non-toxic freezing gel is isolated within the walls of the cooler, guaranteeing that any labels or marks on the vials or tube are not damaged. The indexed and transparent polycarbonate lid shows tube location and matches locations printed on the cooler. ThereMore Product Information