Holder for chromatography column, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F182020000
BELAF182020000EA 2470 DKK
BELAF182020000
Holder for chromatography column, SP Bel-Art
Vagtsøjleholdere
Securely holds as many as eight chromatography columns up to 30 mm (1³/₁₆") in diameter in or between its flexible loops.
  • Rigid plastic construction
  • Easy to secure
  • Can stand alone or be wall-mounted
The rigid plastic back is mounted on two polyethylene support blocks which slide over support stand rods. Tighten the screws to mount the rack at the desired height. Requires two support rods and stands (H18303-0000, sold separately) or can be wall-mounted by removing the support blocks from the back of the unit.