Experimentation kits, bacterial culture and growth study
Leverandør: G-Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BE-201
BE-201EA 3000 DKK
BE-201
Experimentation kits, bacterial culture and growth study
Undervisningsmateriale Biologisk undervisningsmateriale Microbiology Learning Activities
This kit teaches aseptic handling techniques and cultivation of bacteria in liquid culture media and on solid phase agar plates. This kit is designed to educate students about the various stages of the bacterial growth cycle, i.e. lag, log or exponential, stationary and decline or death phases. The kit also teaches the importance of growing bacteria on solid phase agar plates to isolate single colonies of bacteria. This lab activity involves preparation of culture medium and solid agar plates.
  • Aseptic techniques and bacteria culture
  • Study growth curve of bacteria in liquid medium