Super Nuova+™ Hotplate Stirrer
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP88857195 SP88857198 SP88850195
BARNSP88857195EA 8440 DKK
BARNSP88857195 BARNSP88857198 BARNSP88850195
Super Nuova+™ Hotplate Stirrer
Omrørere Magnetomrørere med varme
Thermo Scientific™ SuperNuova+™ stirring hotplate performs with dependable safety and simple operation, units are available in two sizes including a multi-position model.
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Ability to set programs (ramp/dwell) as well as save settings
  • Optional PT1000 temperature probe and splash guard protection shield
Stir trac features for slow-speed stirring, consistent speed control, and stronger magnetic coupling. HotMore Product Information