Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88882016
BARN88882016EA 14020 DKK
BARN88882016
Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator
Rystere Roterende og vuggende rysteborde
The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.
  • Digital display of speed and time
  • Adjustable rocking angle: 95 - 180°
  • Timer range: 0 - 9999 mins
  • Optional multiple size drums offering different capacity of tube rotation