Methyl-4-fluoro-3-hydroxybenzoate ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 4-fluoro-3-hydroxybenzoate

B25683.14 B25683.06
B25683.14EA 2680 DKK
B25683.14 B25683.06
Methyl-4-fluoro-3-hydroxybenzoate ≥98%
Methyl-4-fluoro-3-hydroxybenzoate

Purity: 98+% CAS No.: 214822-96-5 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 89-91° MDL No.: MFCD06797457

Formel: C₈H₇FO₃
Smeltepunkt: 89…91 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06797457
CAS-nummer: 214822-96-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR