2,4-Dichlor-5-methoxyanilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine

B25497.06 B25497.03 B25497.14
B25497.06EA 760 DKK
B25497.06 B25497.03 B25497.14
2,4-Dichlor-5-methoxyanilin ≥98%
2,4-Dichlor-5-methoxyanilin

Purity: 98% CAS No.: 98446-49-2 Molecular Formula: C7H7Cl2NO Formula Weight: 192.05 UN No.: 2810 MDL No.: MFCD00974410

Formel: C₇H₇Cl₂NO
Molekylvægt: 192.04 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00974410
CAS-nummer: 98446-49-2
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR