3-Bromthioanisol ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Bromophenyl methyl sulphide, 1-Bromo-3-(methylthio)benzene

B25406.06
B25406.06EA 2830 DKK
B25406.06
3-Bromthioanisol ≥98%
3-Bromthioanisol

Purity: 98% CAS No.: 33733-73-2 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Refractive Index: 1.6240 Boiling Point: 124-125°/10mm Beilstein Reference: 2078598 MDL No.: MFCD00041395

Formel: C₇H₇BrS
Molekylvægt: 203.1 g/mol
Kogepunkt: 124…125 °C (10 mmHg)
Massefylde: 1.510
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041395
CAS-nummer: 33733-73-2
UN nummer: 3334
ADR: 9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR