(2-Methoxyphenyl)acetonitril ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methoxybenzyl cyanide

B24400.0B B24400.18 B24400.30
B24400.0BEA 5970 DKK
B24400.0B B24400.18 B24400.30
(2-Methoxyphenyl)acetonitril ≥98%
(2-Methoxyphenyl)acetonitril

Purity: 98% CAS No.: 7035-03-2 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 129°(264°F) Melting Point: 66-70° Boiling Point: 141-143°/15mm Beilstein Reference: 2087748 UN No.: 3439 MDL No.: MFCD00001902

Formel: C₉H₉NO
Molekylvægt: 147.18 g/mol
Kogepunkt: 141…143 °C (15 mmHg)
Smeltepunkt: 66…70 °C
Flammepunkt: 129 °C (264 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001902
CAS-nummer: 7035-03-2
EINECS: 230-314-5
UN nummer: 3439
ADR: 6.1 III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR