N-Acetyl-L-methionine ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH

B24312.06 B24312.14
B24312.06EA 619 DKK
B24312.06 B24312.14
N-Acetyl-L-methionine ≥99%
N-Acetyl-L-methionine

Purity: 99% CAS No.: 65-82-7 Molecular Formula: C7H13NO3S Formula Weight: 191.25 Melting Point: 103-107° Beilstein Reference: 1725552 Merck Reference: 14,96 Optical Rotation: -21° (c=1.3 in water) MDL No.: MFCD00064441

Formel: C₇H₁₃NO₃S
Smeltepunkt: 103…107 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064441
CAS-nummer: 65-82-7
EINECS: 200-617-7
Merck Index: 14,96

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR