3-(Brommethyl)heptan 96% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

Stabilization: Stabilised with copper

B23532.14 B23532.22
B23532.14EA 397 DKK
B23532.14 B23532.22
3-(Brommethyl)heptan 96% stabilised
2-Ethylhexyl bromide

Stabilized with copper


Purity: 96% CAS No.: 18908-66-2 Molecular Formula: C8H17Br Formula Weight: 193.13 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Refractive Index: 1.4538 Boiling Point: 94-95°/35mm Beilstein Reference: 1098313 MDL No.: MFCD00000220

Formel: C₈H₁₇Br
Kogepunkt: 94…95 °C (35 mmHg)
Massefylde: 1,086
Flammepunkt: 69 °C (156 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000220
CAS-nummer: 18908-66-2
EINECS: 242-659-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR