α,α,α-Trifluor-o-xylen ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methylbenzotrifluoride, 1-Methyl-2-(trifluoromethyl)benzene, 2-(Trifluoromethyl)toulene

B23453.03 B23453.06
B23453.03EA 516 DKK
B23453.03 B23453.06
α,α,α-Trifluor-o-xylen ≥97%
α,α,α-Trifluor-o-xylen

Purity: 97% CAS No.: 13630-19-8 Molecular Formula: C8H7F3 Formula Weight: 160.14 Refractive Index: 1.4350 Boiling Point: 125-126° UN No.: 1993 MDL No.: MFCD00153204

Formel: C₈H₇F₃
Kogepunkt: 125…126 °C
Massefylde: 1,174
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00153204
CAS-nummer: 13630-19-8
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR