N-Isopropylbenzylamin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Benzyl-N-isopropylamine, N-Benzylisopropylamine

B23243.22 B23243.36
B23243.22EA 706 DKK
B23243.22 B23243.36
N-Isopropylbenzylamin 97%
N-Isopropylbenzylamin

Purity: 97% CAS No.: 102-97-6 Molecular Formula: C10H15N Formula Weight: 149.24 Flash Point: 87°(190°F) Refractive Index: 1.5020 Boiling Point: 199-201° MDL No.: MFCD00008863

Formel: C₆H₅CH₂N(CH₃)(CH₂CH₃)
Molekylvægt: 149.24 g/mol
Kogepunkt: 199…201 °C
Smeltepunkt: ∼ 0 °C
Massefylde: 0.910
Flammepunkt: 87 °C (190 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008863
CAS-nummer: 102-97-6
EINECS: 203-067-6
UN nummer: 2735
ADR: 8 III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR