4-Nitrobenzylamine hydrochloride 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Nitrophenyl)methanamine hydrochloride

B21748.03 B21748.06 B21748.14
B21748.03EA 549 DKK
B21748.03 B21748.06 B21748.14
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride

Purity: 97% CAS No.: 18600-42-5 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: 255-257° dec. Beilstein Reference: 3629994

Formel: C₇H₉ClN₂O₂
Smeltepunkt: 255…257 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012863
CAS-nummer: 18600-42-5
EINECS: 242-441-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR