1-Bromo-4-(trifluoromethylthio)benzene ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Trifluoromethylthio)bromobenzene, 4-Bromophenyl trifluoromethyl sulphide

B20874.03 B20874.06
B20874.03EA 443 DKK
B20874.03 B20874.06
1-Bromo-4-(trifluoromethylthio)benzene ≥97%
1-Bromo-4-(trifluoromethylthio)benzene

Purity: 97% CAS No.: 333-47-1 Molecular Formula: C7H4BrF3S Formula Weight: 257.07 Flash Point: 83°(181°F) Refractive Index: 1.5170 Boiling Point: 199-200° Beilstein Reference: 2361926 MDL No.: MFCD00040840

Formel: C₇H₄BrF₃S
Kogepunkt: 199…200 °C
Massefylde: 1,710
Flammepunkt: 83 °C (181 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040840
CAS-nummer: 333-47-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR