Reagent reservoirs, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RES-V-50-SI RES-2CV-25 RES-2CV-25-S RES-2CV-25-SI RES-V-100 RES-V-100-S RES-V-100-SI RES-V-25 RES-V-25-S RES-V-25-SI RES-V-50-S RES-V-50
AXYGRES-V-50-SIEA 1900 DKK
AXYGRES-V-50-SI AXYGRES-2CV-25 AXYGRES-2CV-25-S AXYGRES-2CV-25-SI AXYGRES-V-100 AXYGRES-V-100-S AXYGRES-V-100-SI AXYGRES-V-25 AXYGRES-V-25-S AXYGRES-V-25-SI AXYGRES-V-50-S AXYGRES-V-50
Reagent reservoirs, Axygen®
Reagens reservoirs
PS, white. These disposable reservoirs are designed to accommodate all multi channel pipettors.
  • Certified RNase-/DNase-free
  • Non pyrogenic
  • Four corners and sharp V-bottom shape ensures maximum sample retrieval
  • Available sterile or non sterile
  • 25 ml reservoir also available with two channels (divided)