Culture media, in Petri dishes, Slanetz Bartley medium
Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
443812.0922
APPC443812.0922EA 1130 DKK
APPC443812.0922
Culture media, in Petri dishes, Slanetz Bartley medium
Mikrobiologiske medier Klargjort medium
Prepared medium for the enumeration of Enterococci.
  • Pinkish beige