Kaliumiodid 99.0-100.5% USP, Ph. Eur., BP, pharma grade

Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dipotassium diiodide, Kali iodide

141542.1209 141542.1210 141542.1211 141542.1214
APPC141542.1209EA 1860 DKK
APPC141542.1209 APPC141542.1210 APPC141542.1211 APPC141542.1214
Kaliumiodid 99.0-100.5% USP, Ph. Eur., BP, pharma grade
Potassium iodide
Formel: Kl
Molekylvægt: 166 g/mol
Kogepunkt: 1330 °C
Smeltepunkt: 680 °C
MDL Number: MFCD00011405
CAS-nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Bestil nu

Specifikationer Testresultater

Assay calc. a.d.s 99.0 - 100.5%
Alkalinity Passes Test
Insoluble matter in H₂O 0.01%
Loss on drying at 105 °C 1.0%
Sulfate (SO₄) 0.015%
Residual Solvents (Ph. Eur./USP) Passes Test
Chloride and Bromide (as Cl) 0.05%
Nitrate, Nitrite and Ammonia Passes Test
Thiosulfate Passes Test
Iodate Passes Test
Heavy metals (as Pb) 0.001%
Ba Passes Test
Fe 0.002%
Cd, Class 1 - (ICH Q3D) 0.5 ppm
Pb, Class 1 - (ICH Q3D) 0.5 ppm
As, Class 1 - (ICH Q3D) 1.5 ppm
Hg, Class 1 - (ICH Q3D) 1.5 ppm
Co, Class 2A - (ICH Q3D) 5 ppm
V, Class 2A - (ICH Q3D) 10 ppm
Ni, Class 2A - (ICH Q3D) 20 ppm
Tl, Class 2B - (ICH Q3D) 0.8 ppm
Au, Class 2B - (ICH Q3D) 10 ppm
Pd, Class 2B - (ICH Q3D) 10 ppm
Ir, Class 2B - (ICH Q3D) 10 ppm
Os, Class 2B - (ICH Q3D) 10 ppm
Rh, Class 2B - (ICH Q3D) 10 ppm
Ru, Class 2B - (ICH Q3D) 10 ppm
Se, Class 2B - (ICH Q3D) 15 ppm
Ag, Class 2B - (ICH Q3D) 15 ppm
Pt, Class 2B - (ICH Q3D) 10 ppm
Li, Class 3 - (ICH Q3D) 55 ppm
Sb, Class 3 - (ICH Q3D) 120 ppm
Ba, Class 3 - (ICH Q3D) 140 ppm
Mo, Class 3 - (ICH Q3D) 25 ppm
Cu, Class 3 - (ICH Q3D) 250 ppm
Sn, Class 3 - (ICH Q3D) 600 ppm
Cr, Class 3 - (ICH Q3D) 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR