Citronsyre monohydrat, crystals USP, JP, Ph. Eur., BP, pure

Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

141018.0416 141018.1211 141018.1214
APPC141018.0416EA 0 DKK
APPC141018.0416 APPC141018.1211 APPC141018.1214
Citronsyre monohydrat, crystals USP, JP, Ph. Eur., BP, pure
Citronsyre monohydrat

Synonyms: 2-Hydroxy-1,2,3-Propanetricarboxilic acid.

Formel: HOC(COOH)(CH₂COOH)₂·H₂O
Smeltepunkt: 135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149972
CAS-nummer: 5949-29-1
EINECS: 201-069-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (Acidim.) calc. a.a.s. 99,5 - 100,5%
Identity according to Pharmacopoeias Passes test
Appearance of solution Passes test
Insoluble matter in H₂O 0,01%
Darkened substances by H₂SO₄ Passes test
Residue on ignition (as SO₄) 0,05%
Chloride (Cl)  0,005%
Sulfate (SO₄)  0,01%
Colour of solution Passes test
Residual solvents (Ph.Eur/USP) Passes test
Oxalate (C₂O₄)  0,035%
Water (H₂O) 7,5 - 8,8%
Heavy metals (as Pb) 0,001%
Barium (Ba) 140 ppm
Calcium (Ca) 0,02%
Magnesium (Mg) 0,005%
Cadmium (Cd) 0,5 ppm
Lead (Pb) 0,5 ppm
Arsenic (As) 1 ppm
Mercury (Hg) 1,5 ppm
Cobalt (Co) 5 ppm
Vanadium (V) 10 ppm
Nickel (Ni) 20 ppm
Thallium (Tl) 5 ppm
Gold (Au) 10 ppm
Palladium (Pd) 10 ppm
Iridium (Ir) 10 ppm
Osmium (Os) 10 ppm
Rhodium (Rh) 10 ppm
Ruthenium (Ru) 10 ppm
Selenium (Se) 15 ppm
Silver (Ag) 15 ppm
Platinum (Pt) 10 ppm
Lithium (Li) 55 ppm
Antimony (Sb) 120 ppm
Molybdenum (Mo) 25 ppm
Copper (Cu) 250 ppm
Tin (Sn) 600 ppm
Chromium (Cr) 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR