2-Brom-4-Fluor-6-(trifluormethyl)anilin 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Bromo-α,α,α,4-tetrafluoro-o-toluidine

PC906319-25G PC906319-5G PC906319-100G
APOSPC906319-25GEA 985 DKK
APOSPC906319-25G APOSPC906319-5G APOSPC906319-100G
2-Brom-4-Fluor-6-(trifluormethyl)anilin 98%
2-Brom-4-Fluor-6-(trifluormethyl)anilin
Formel: C₇H₄BrF₄N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04973756
CAS-nummer: 875664-27-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR