Yttrium(III)fluorid 99.99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Yttrium trifluoride, Yttrium(III) fluoride

PC8024-100G PC8024-25G
APOSPC8024-100GEA 2850 DKK
APOSPC8024-100G APOSPC8024-25G
Yttrium(III)fluorid 99.99%
Yttrium(III) fluoride
Formel: F₃Y
Smeltepunkt: 1387 °C
Massefylde: 4,01 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00049714
CAS-nummer: 13709-49-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR