3-Chlor-5-(trifluormethyl)anisol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC8005-5G PC8005-1G
APOSPC8005-5GEA 1290 DKK
APOSPC8005-5G APOSPC8005-1G
3-Chlor-5-(trifluormethyl)anisol 98%
3-Chlor-5-(trifluormethyl)anisol
Formel: C₈H₆ClF₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115855
CAS-nummer: 886497-07-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR