3-Chlor-5-(trifluormethyl)phenol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Chloro-5-hydroxybenzotrifluoride

PC8004-1G PC8004-25G PC8004-5G
APOSPC8004-1GEA 546 DKK
APOSPC8004-1G APOSPC8004-25G APOSPC8004-5G
3-Chlor-5-(trifluormethyl)phenol
3-Chlor-5-(trifluormethyl)phenol
Formel: C₇H₄ClF₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115852
CAS-nummer: 570391-18-3
UN nummer: 3265
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR