Tris[4-(trifluoromethyl)phenyl]phosphine 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine

PC7919G-1G PC7919G-5G PC7919G-10G
APOSPC7919G-1GEA 273 DKK
APOSPC7919G-1G APOSPC7919G-5G APOSPC7919G-10G
Tris[4-(trifluoromethyl)phenyl]phosphine 95+%
Tris[4-(trifluoromethyl)phenyl]phosphine
Formel: C₂₁H₁₂F₉P
Smeltepunkt: 73…76 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00058883
CAS-nummer: 13406-29-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR