Perfluoro(methyldecalin) 85%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Heptadecafluorodecahydro(trifluoromethyl)naphthalene, Perfluoro(methyldecahydronaphthalene)

PC6120-25G
APOSPC6120-25GEA 121 DKK
APOSPC6120-25G
Perfluoro(methyldecalin) 85%
Perfluoro(methyldecalin)
Formel: C₁₁F₂₀
Molekylvægt: 512.09 g/mol
Kogepunkt: 160 °C
Massefylde: 1.96 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014326
CAS-nummer: 306-92-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR