4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520672-1G PC520672-5G PC520672-250MG
APOSPC520672-1GEA 1110 DKK
APOSPC520672-1G APOSPC520672-5G APOSPC520672-250MG
4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 97%
4-Methyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Formel: C₇H₅F₃N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11100557
CAS-nummer: 306960-74-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR