5-Chloro-2-methylbenzotrifluoride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520576-10G PC520576-1G PC520576-5G
APOSPC520576-10GEA 7780 DKK
APOSPC520576-10G APOSPC520576-1G APOSPC520576-5G
5-Chloro-2-methylbenzotrifluoride 97%
5-Chloro-2-methylbenzotrifluoride
Formel: C₈H₆ClF₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09037961
CAS-nummer: 1839-92-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR