2-Brom-4-(trifluormethyl)phenol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Bromo-4-hydroxybenzotrifluoride

PC49531-5G PC49531-25G PC49531-100G
APOSPC49531-5GEA 164 DKK
APOSPC49531-5G APOSPC49531-25G APOSPC49531-100G
2-Brom-4-(trifluormethyl)phenol
2-Brom-4-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06657954
CAS-nummer: 81107-97-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR